

2,2-Difluoro-1,3-benzodioxole ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1583-59-1
:
Formula:C₇H₄F₂O₂
MDL Number:MFCD00236217
:158.1 g/mol
Boiling Pt:129…130 °C
Density:1.308
Flash Pt:32 °C (90 °F)
Storage Temperature:Ambient
EINECS:216-431-4
UN:1993
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together