
7-Isopropyl-1,4-dimethylazulene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:489-84-9
:
Formula:C₁₅H₁₈
MDL Number:MFCD00003811
:198.31 g/mol
Merck Index:12,04581
Boiling Pt:153 °C (7Torr)
Melting Pt:29…31 °C
Density:0.97 g/cm³
Flash Pt:>110 °C
EINECS:207-701-2
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Dark blue Low melting crystalline solid
- Infrared spectrum:Conforms
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together