ContactUsLinkComponent
N-Methyl-2-pyrrolidone (NMP), extra dry over Molecular Sieve 99.5%, AcroSeal®
Irritant
Health
N-Methyl-2-pyrrolidone (NMP), extra dry over Molecular Sieve 99.5%, AcroSeal®
Certificates
undefined
Irritant
Health
N-Methyl-2-pyrrolidone (NMP), extra dry over Molecular Sieve 99.5%, AcroSeal®
Certificates
Specification Test Results
  • Appearance:
    Clear liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 % (Coulometric)
  • GC:
    ≥99.5 %
  • Color scale:
    ≤50 APHA