Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
Piperidine, EMSURE® for analysis, Supelco®
  1.09724.0500
 :  Merck
 :  
 
 
 
Piperidine, EMSURE® for analysis, Supelco®
  1.09724.0500
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Solubility:
    (20 °C) miscible in all proportions
  • Melting Point:
    -10 °C
  • Molar Mass:
    85.15 g/mol
  • Boiling Point:
    106 °C (1013 hPa)
  • Vapor Pressure:
    34 hPa (20 °C)
  • Flash Point:
    16 °C
  • Explosion Limit:
    1.5 - 10.3 %(V)
  • Density:
    0.86 g/cm3 (20 °C)
  • pH:
    12.6 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    320 °C

 

 

 :
 :85.15 g/mol
 :12,07621
 :107.3 °C (1013 hPa)
 :–10 °C
 :0.8622 g/cm³ (20 °C)
 :16 °C
 :Ambient
 :203-813-0
 :2401
 :8 (3),
 :M-Clarity™ quality level = MQ300
 :GHS02
 :GHS06
 :GHS05
 :110-89-4
 :C₅H₁₁N
 :MFCD00005979