Piperidin ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-89-4
:
Formula:C₅H₁₁N
MDL Number:MFCD00005979
Merck Index:14,7468
Boiling Pt:105…106 °C
Melting Pt:–11 °C
Density:0,862
Flash Pt:3 °C (37 °F)
Storage Temperature:Ambient
EINECS:203-813-0
UN:2401
ADR:8,I
:GHS02
:GHS06
:GHS05
Related Information
Frequently Bought Together