2-Methyl-3-butyne-2-ol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:115-19-5
:
Formula:C₅H₈O
MDL Number:MFCD00004467
Merck Index:12,06113
Boiling Pt:104 °C (1013 hPa)
Melting Pt:3 °C
Density:0.86 g/cm³ (20 °C)
Flash Pt:20 °C
Storage Temperature:Ambient
EINECS:204-070-5
UN:1993
ADR:3,II
:M-Clarity™ quality level = MQ200
:84.12 g/mol
Frequently Bought Together