


2-Methyl-3-butyne-2-ol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:115-19-5
:
Formula:C₅H₈O
MDL Number:MFCD00004467
Merck Index:12,06113
Boiling Pt:104 °C (760 mmHg)
Melting Pt:3 °C
Density:0.8608 g/cm³
Flash Pt:21 °C
Storage Temperature:Ambient
UN:1987
ADR:3,II
:84.12 g/mol
EINECS:204-070-5
:GHS05
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4190 to 1.4220 (20°C, 589 nm)
- GC:≥97.5 %
- Color scale:≤100 APHA
- Iron (Fe):≤0.001 %
- Magnesium (Mg):≤0.05 %
- Potassium (K):≤0.01 %
- Sodium (Na):≤0.02 %
- Strontium (Sr):≤0.05 %
- Heavy metals (as Pb):≤0.005 %
- Titrable acid:≤0.035 meq/g
- Insoluble matter:≤0.005 %
Specification Test Results
Frequently Bought Together