
5-Hexyn-1-ol
Catalog # ABCRA11071.0025
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
About this item
:
:98.14 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:928-90-5
Formula:C₆H₁₀O
MDL Number:MFCD00002980