

3-Butyne-1-ol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:927-74-2
:
Formula:C₄H₆O
MDL Number:MFCD00002955
:70.09 g/mol
Boiling Pt:128.8 °C (1013 hPa)
Melting Pt:–64 °C
Density:0.9262 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:213-161-9
ADR:3,III
:M-Clarity™ quality level = MQ200
UN:1987
:GHS02
:GHS07
Frequently Bought Together