3-Hexyn-2-ol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:109-50-2
:
Formula:C₆H₁₀O
MDL Number:MFCD00041586
:98.14 g/mol
Boiling Pt:80 °C (1013 hPa)
Density:0.8786 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-676-7
UN:1987
ADR:3,III
:M-Clarity™ quality level = MQ100
Frequently Bought Together