
3-Hexyn-1-ol, Sigma-Aldrich®
Catalog # SIAL244430-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:98.14 g/mol
Boiling Pt:159 °C (1013 hPa)
Melting Pt:42 °C
Density:0.902 g/cm³ (14 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:213-685-8
:GHS07
CAS Number:1002-28-4
Formula:C₆H₁₀O
MDL Number:MFCD00002957