Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Hexyn-1-ol
2-Hexyn-1-ol, Sigma-Aldrich®
2-Hexyn-1-ol, Sigma-Aldrich®
SIAL630829-5G
:
Merck
:
2-Hexyn-1-ol, Sigma-Aldrich®
SIAL630829-5G
:
Merck
:
Typ balení:
Glass bottle for solids
Bal.:
5 g
Sekvence:
3173
:
:
M-Clarity™ quality level = MQ100
:
212-128-6
:
764-60-3
:
C₆H₁₀O
:
MFCD00039540