
trans-2-Hydroxycinnamic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:614-60-8
:
Formula:C₉H₈O₃
MDL Number:MFCD00004379
:164.16 g/mol
:M-Clarity™ quality level = MQ100
EINECS:210-386-4
:GHS07
Frequently Bought Together