trans-3-Hexene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13269-52-8
:
Formula:C₆H₁₂
MDL Number:MFCD00009386
:84.16 g/mol
Boiling Pt:67 °C (1013 hPa)
Melting Pt:–115.4 °C
Density:0.6772 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:236-261-4
ADR:3,II
:M-Clarity™ quality level = MQ100
UN:3295
Frequently Bought Together