
cis-2-Butene-1,4-diol 96% remainder trans-isomer
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6117-80-2
:
Formula:C₄H₈O₂
MDL Number:MFCD00002924
:88.11 g/mol
Merck Index:12,02390
Boiling Pt:131…132 °C (12 mmHg)
Melting Pt:6…8 °C
Density:1.070
Flash Pt:127 °C (260 °F)
EINECS:228-085-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together