

cis-3-Hexen-1-ol 98%
:928-96-1
:
:C₆H₁₂O
:MFCD00063217
:100.16 g/mol
:13,04719
:156…157 °C (760 mmHg)
:–60 °C
:0.848 g/cm³
:59 °C
:Ambient
:213-192-8
:1987
:3,III
:GHS02
:GHS07
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4380 to 1.4410 (20°C, 589 nm)
- Density:(20°C) 0.845 to 0.853 g/ml
- GC:≥97.5 %
Frequently Bought Together