Order Entry
Czech Republic
ContactUsLinkComponent
 
10-Undecen-1-ol 98+% stabilised
 
undefined
10-Undecen-1-ol 98+% stabilised

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4480 to 1.4530 (20°C, 589 nm)
  • GC:
    ≥98.0 %
  • Stabilizer:
    Abt 0.1% alpha-tocopherol (synthetic)

 

 

Frequently Bought Together