1H,1H,2H-Perfluoro-1-hexene, Sigma-Aldrich®
Catalog # SIAL371459-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:246.08 g/mol
Boiling Pt:59…60 °C (1013 hPa)
Density:1.41 g/cm³ (20 °C)
ADR:3,II
:M-Clarity™ quality level = MQ100
EINECS:243-053-7
UN:1993
CAS Number:19430-93-4
Formula:C₆H₃F₉
MDL Number:MFCD00042338