


Dimethylmaleát 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:624-48-6
:
Formula:C₆H₈O₄
MDL Number:MFCD00008459
Boiling Pt:204…205 °C
Melting Pt:–19 °C
Density:1,151
Flash Pt:91 °C (195 °F)
EINECS:210-848-5
UN:2810
ADR:6.1,III
:GHS06
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together