Methyl oleate, Technical Grade C18 71-90%, C18:1 >65% of C18
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-62-9
:67762-38-3
Formula:C₁₉H₃₆O₂
MDL Number:MFCD00009578
:296.49 g/mol
Merck Index:14,6828
Boiling Pt:168…170 °C (2 mmHg)
Melting Pt:–20 °C
Density:0.876
Flash Pt:163 °C (325 °F)
Storage Temperature:Ambient
EINECS:203-992-5
Related Information
Frequently Bought Together