Methyl oleate, technical
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67762-38-3
Formula:C₁₉H₃₆O₂
MDL Number:MFCD00009578
:296.49 g/mol
Merck Index:12,06965
Boiling Pt:186 °C (9 mmHg)
Melting Pt:–20 °C
Density:0.87 g/cm³
Flash Pt:170 °C
Storage Temperature:Refrigerator
EINECS:203-992-5
Specification Test Results
- Appearance:Clear yellow to amber Viscous liquid
- Infrared spectrum:Conforms
- GC:≥55 %
- HPLC:≥98.0 %
Specification Test Results
Frequently Bought Together