Ethyl oleate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester
CAS Number:111-62-6
:85049-36-1
Formula:C₂₀H₃₈O₂
MDL Number:MFCD00009579
:310.52 g/mol
Merck Index:12,06965
Boiling Pt:386 °C (1013 hPa)
Melting Pt:–32 °C
Density:0.87 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:174 °C
Storage Temperature:Ambient
EINECS:203-889-5
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together