Ethyl oleate 70%, tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-62-6
:85049-36-1
Formula:C₂₀H₃₈O₂
MDL Number:MFCD00009579
:310.52 g/mol
Merck Index:12,06965
Boiling Pt:205…208 °C
Melting Pt:–32 °C
Density:0.87
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:203-889-5
Related Information
Frequently Bought Together