Order Entry
Czech Republic
ContactUsLinkComponent
Acrylamide, 4x crystallised
Toxic
Health
Acrylamide, 4x crystallised
Catalog # A1090.0500
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Toxic
Health
Acrylamide, 4x crystallised
Catalog # A1090.0500
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :71.08 g/mol
Boiling Pt:232 °C (1013 hPa)
Melting Pt:84.5 °C
Density:1.122 g/cm³ (20 °C)
Flash Pt:138 °C
Storage Temperature:Refrigerator
EINECS:201-173-7
UN:2074
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:79-06-1
Formula:C₃H₅NO
MDL Number:MFCD00008032