

Fumaroyl chloride 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:627-63-4
:
Formula:C₄H₂Cl₂O₂
MDL Number:MFCD00000733
:152.96 g/mol
Boiling Pt:161…164 °C (760 mmHg)
Melting Pt:–2 °C
Density:1.41 g/cm³
Flash Pt:73 °C
Storage Temperature:Refrigerator
EINECS:211-005-4
UN:1780
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4980 to 1.5010 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
Frequently Bought Together