

Tetrachloroethylene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:127-18-4
:
Formula:C₂Cl₄
MDL Number:MFCD00000834
:165.83 g/mol
Merck Index:12,09332
Boiling Pt:121.1 °C (1013 hPa)
Melting Pt:–22 °C
Density:1.63 g/cm³ (20 °C)
UN:1897
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:204-825-9
:GHS08
:GHS09
Storage Temperature:Ambient
Frequently Bought Together