



cis-1,3-Dichloropropene, Sigma-Aldrich®
Catalog # SIAL377414-1ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:110.97 g/mol
Boiling Pt:104.3 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.225 g/cm³ (20 °C)
EINECS:233-195-8
UN:2047
ADR:3,II
:M-Clarity™ quality level = MQ100
:GHS02
:GHS06
:GHS08
:GHS09
CAS Number:10061-01-5
Formula:C₃H₄Cl₂
MDL Number:MFCD00062949