Periodic acid 0,5% in aqueous solution, Sigma-Aldrich®
Catalog # 1.00482.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Nuclei:Blue
- polysaccharides:purple
- glycogen:purple
- neutral mucopolysaccharides:purple
- mucoproteins:purple
- glycoproteins:purple
- glycolipids:purple
- phospholipids:purple
- Basement membrane:purple
- Collagen:purple
Specification Test Results
About this item
:
:227.94 g/mol
Storage Temperature:Ambient
UN:3264
ADR:8,III
:1
CAS Number:10450-60-9
Formula:H₅IO₆
MDL Number:MFCD00011344