
trans-2-Nonenal ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18829-56-6
:
Formula:C₉H₁₆O
MDL Number:MFCD00007012
:140.23 g/mol
Merck Index:14,6676
Boiling Pt:98…100 °C (12 mmHg)
Density:0.846
Flash Pt:84 °C (184 °F)
EINECS:242-609-6
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together