Order Entry
Czech Republic
ContactUsLinkComponent
4-Bromo-1,1,2-trifluoro-1-butene, Sigma-Aldrich®
4-Bromo-1,1,2-trifluoro-1-butene, Sigma-Aldrich®
Catalog # SIAL443743-5ML
Supplier:  Merck
CAS Number:  
4-Bromo-1,1,2-trifluoro-1-butene, Sigma-Aldrich®
Catalog # SIAL443743-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
Boiling Pt:95 °C (1013 hPa)
Density:1.639 g/cm³ (20 °C)
EINECS:234-019-2
ADR:3,II
 :M-Clarity™ quality level = MQ100
 :188.97 g/mol
UN:1993
CAS Number:10493-44-4
Formula:C₄H₄BrF₃
MDL Number:MFCD00039274