Order Entry
Czech Republic
ContactUsLinkComponent
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) ≥99.5% suitable for buffer solutions
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) ≥99.5% suitable for buffer solutions
Supplier:  PanReac AppliChem
Certificates
undefined
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) ≥99.5% suitable for buffer solutions
Supplier:  PanReac AppliChem

Specification Test Results

  • Assay (titr.):
    min. 99.5 %
  • pH (1 %; H2O):
    4.7 - 6.0
  • Heavy metals (as Pb):
    max. 0.001 %
  • Water:
    max. 0.5 %
  • Chloride:
    max. 0.05 %
  • Sulfate:
    max. 0.05 %

Specification Test Results

Frequently Bought Together