Weighing boats, aluminium
SIALW1126-100EA
: Merck
- W×D×H:12 x 4 x 4 mm mm
- Pk:100
Made of aluminium.
- Lightweight
- Good chemical resistance
- Good temperature resistance
Ideal for electrobalance weighing and combustion analysis. Boats are rectangular with flat bottom and tweezer-grip tab for easy handling.