



2,2-Dimethylhexane, Sigma-Aldrich®
Catalog # SIAL111457-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:114.23 g/mol
Boiling Pt:107 °C (1013 hPa)
Melting Pt:–91 °C
Density:0.6955 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-689-4
UN:1262
ADR:3,II
:M-Clarity™ quality level = MQ100
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:590-73-8
Formula:C₈H₁₈
MDL Number:MFCD00009485