
2,5-Dimethyl-2,5-hexanediol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-03-2
:
Formula:C₈H₁₈O₂
MDL Number:MFCD00004473
:146.23 g/mol
Boiling Pt:214 °C (1013 hPa)
Melting Pt:85…88 °C
Density:0.898 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:203-731-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together