5-Methyl-1-hexanol, Sigma-Aldrich®
Catalog # SIAL493953-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:116.2 g/mol
Boiling Pt:172 °C (1013 hPa)
Melting Pt:–59 °C
Density:0.8192 g/cm³ (24 °C)
EINECS:211-023-2
UN:1987
ADR:3,III
:M-Clarity™ quality level = MQ100
CAS Number:627-98-5
Formula:C₇H₁₆O
MDL Number:MFCD00072703