

Isoamyl alcohol, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:123-51-3
:
Formula:(CH₃)₂CHCH₂CH₂OH
MDL Number:MFCD00002934
:88.15 g/mol
Merck Index:13,05214
Boiling Pt:130 °C (1013 hPa)
Melting Pt:–117 °C
Density:0.812 g/cm³ (20 °C)
EINECS:204-633-5
UN:1105
ADR:3,III
:M-Clarity™ quality level = MQ300
Flash Pt:42 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Solubility:25 g/l (20 °C)
- Melting Point:-117 °C
- Molar Mass:88.15 g/mol
- Boiling Point:131 °C (1013 hPa)
- Vapor Pressure:3 hPa (20 °C)
- Flash Point:45 °C
- Explosion Limit:1.2 - 8 %(V)
- Density:0.81 g/cm3 (20 °C)
- pH:7 (25 g/l, H2O, 20 °C)
- Ignition Point:335 °C
Specification Test Results
Related Information
Frequently Bought Together