3,7-Dimethyl-1-octanol, Sigma-Aldrich®
Catalog # SIALW239100-8KG-K
Supplier: Merck
CAS Number:
Specifications
- Pk:8 kg
- Seq:4138
Specifications
About this item
:
:158.28 g/mol
Boiling Pt:95 °C (1013 hPa)
Density:0.849 g/cm³ (0 °C)
:M-Clarity™ quality level = MQ300
EINECS:203-374-5
CAS Number:106-21-8
Formula:C₁₀H₂₂O
MDL Number:MFCD00002936