Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
tert-Butanol, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
  1.09629.0500
 :  Merck
   Safer Human and Environmental Health
 :  
undefined
 
 
tert-Butanol, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
  1.09629.0500
 :  Merck
   Safer Human and Environmental Health
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    24 - 25 °C
  • Molar Mass:
    74.12 g/mol
  • Boiling Point:
    81 - 83 °C (1013 hPa)
  • Vapor Pressure:
    40.7 hPa (20 °C)
  • Flash Point:
    14 °C
  • Explosion Limit:
    2.3 - 8.0 %(V)
  • Density:
    0.78 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) neutral
  • Ignition Point:
    490 °C

 

 

 :
 :74.12 g/mol
 :12,01577
 :82.2 °C (1013 hPa)
 :25.3 °C
 :0.7887 g/cm³ (20 °C)
 :11 °C
 :200-889-7
 :1120
 :3,
 :M-Clarity™ quality level = MQ300
 :GHS02
 :GHS07
 :Ambient
 :75-65-0
 :(CH₃)₃COH
 :MFCD00004464