

2-Methyl-2-butanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-85-4
:
Formula:C₅H₁₂O
MDL Number:MFCD00004478
:88.15 g/mol
Merck Index:13,07218
Boiling Pt:102 °C (1013 hPa)
Melting Pt:–8.4 °C
Density:0.806 g/cm³ (20 °C)
EINECS:200-908-9
UN:1105
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:20 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together