

2-Methyl-2-butanol
Catalog # EHERC15084410
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:88.15 g/mol
Merck Index:13,07218
Boiling Pt:102 °C (1013 hPa)
Melting Pt:–8.4 °C
Density:0.806 g/cm³ (20 °C)
Flash Pt:20 °C
Storage Temperature:Ambient
EINECS:200-908-9
UN:1105
ADR:3,II
:GHS02
:GHS07
CAS Number:75-85-4
Formula:C₅H₁₂O
MDL Number:MFCD00004478