Order Entry
Czech Republic
ContactUsLinkComponent
2,3-Dimethyl-2-butanol 98%
Flammable
2,3-Dimethyl-2-butanol 98%
Certificates
undefined
Flammable
2,3-Dimethyl-2-butanol 98%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4160 to 1.4180 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together