AMP (2-Amino-2-methylpropanol), Sigma-Aldrich®
: Merck
: 2-Amino-2-methyl-1-propanol, Isobutanolamine, AMP
:124-68-5
:
:(CH₃)₂C(NH₂)CH₂OH
:MFCD00008051
:165
:18…26 °C
:0.667 g/cm³ (20 °C)
:89.14 g/mol
:67
:13,00448
:M-Clarity™ quality level = MQ200
:204-709-8
:GHS07
:Ambient
- Environmentally Preferable:
Frequently Bought Together