

Isopropyl isothiocyanate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2253-73-8
:
Formula:C₄H₇NS
MDL Number:MFCD00037830
:101.17 g/mol
Boiling Pt:136…137 °C (1013 hPa)
Density:0.9475 g/cm³ (20 °C)
EINECS:218-851-3
UN:1993
ADR:3,III
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
Frequently Bought Together