1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, Sigma-Aldrich®
Catalog # SIAL469548-5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:196.17 g/mol
Boiling Pt:178 °C (1013 hPa)
Density:1.129 g/cm³ (20 °C)
ADR:3,III
:M-Clarity™ quality level = MQ200
EINECS:245-211-0
UN:1224
CAS Number:22767-90-4
Formula:C₈H₁₁F₃O₂
MDL Number:MFCD00040841