Order Entry
Czech Republic
ContactUsLinkComponent
 
L(-)-Glutathione (reduced form) ≥98%, off-white powder
 
L(-)-Glutathione (reduced form) ≥98%, off-white powder

 

  • Identity Test A:
    Passes
  • Identity Test B:
    Passes
  • Purity:
    ≥98%
  • FTIR:
    Conforms to Standard
  • Optical Rotation:
    (−)16 ±5° (c=2, water)
  • pH:
    Acidic (3,5% aq soln)

 

Frequently Bought Together