

2,6-Dimethyl-4-heptanone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-83-8
:
Formula:C₉H₁₈O
MDL Number:MFCD00008940
:142.24 g/mol
Boiling Pt:165 °C (1013 hPa)
Melting Pt:–46 °C
Density:0.806 g/cm³ (20 °C)
Flash Pt:60 °C
UN:1224
ADR:3,III
:M-Clarity™ quality level = MQ300
EINECS:203-620-1
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together