
n-Dodecane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:112-40-3
:
Formula:H₃C(CH₂)₁₀CH₃
MDL Number:MFCD00008969
:170.34 g/mol
Boiling Pt:216 °C (1013 hPa)
Melting Pt:–9.6 °C
Density:0.7487 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:74 °C
EINECS:203-967-9
:GHS08
Storage Temperature:Ambient
Frequently Bought Together