ContactUsLinkComponent
1-Octanesulphonic acid sodium salt for HPLC
Irritant
1-Octanesulphonic acid sodium salt for HPLC
Certificates
undefined
Irritant
1-Octanesulphonic acid sodium salt for HPLC
Certificates
Specification Test Results
  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤2 %
  • UV:
    at 250 nm A: ≤0.05
  • Titration after Ion exchange:
    ≥98.0 %