ContactUsLinkComponent
1-Hexanesulphonic acid sodium salt ≥98.0% for HPLC, for ion pair chromatography, J.T.Baker®
1-Hexanesulphonic acid sodium salt ≥98.0% for HPLC, for ion pair chromatography, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
1-Hexanesulphonic acid sodium salt ≥98.0% for HPLC, for ion pair chromatography, J.T.Baker®
Supplier:  Avantor
Certificates
Specification Test Results
  • For Laboratory,Research,or Manufacturing Use:
    = 98.0 %
  • Assay (as C₆H₁₃SO₃Na) (by acidimetry):
    ≥98.0 %
  • UV Absorbance of 0.25 M Solution - 250 nm:
    ≤0.050
  • UV Absorbance of 0.25 M Solution - 240 nm:
    ≤0.050
  • UV Absorbance of 0.25 M Solution - 230 nm:
    ≤0.050
  • UV Absorbance of 0.25 M Solution - 220 nm:
    ≤0.060
  • UV Absorbance of 0.25 M Solution - 210 nm:
    ≤0.080
  • UV Absorbance of 0.25 M Solution - 200 nm:
    ≤0.200