
1-Butanesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2386-60-9
:
Formula:C₄H₉ClO₂S
MDL Number:MFCD00007464
:156.63 g/mol
Boiling Pt:80…81 °C (9 mmHg)
Melting Pt:–29 °C
Density:1.210
Flash Pt:79 °C (174 °F)
Storage Temperature:Ambient
EINECS:219-204-8
UN:3265
ADR:8,II
:GHS05
Related Information
Frequently Bought Together